Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate, tech

Lieferant: Apollo Scientific

PC28154-1G
APOSPC28154-1GEA 1570 EUR
APOSPC28154-1G
Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate, tech
Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate
Formel: C₁₅H₁₁FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04126009
CAS-Nummer: 866133-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR