2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylic acid, tech

Lieferant: Apollo Scientific

PC28155-1G
APOSPC28155-1GEA 121 EUR
APOSPC28155-1G
2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylic acid, tech
2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylic acid
Formel: C₁₄H₉FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03848674
CAS-Nummer: 866133-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR