N-[2-(4-Methylpiperazin-1-yl)-5-(trifluoromethyl)phenyl]hydroxylamine, tech

Lieferant: Apollo Scientific

PC28156-1G
APOSPC28156-1GEA 1570 EUR
APOSPC28156-1G
N-[2-(4-Methylpiperazin-1-yl)-5-(trifluoromethyl)phenyl]hydroxylamine, tech
N-[2-(4-Methylpiperazin-1-yl)-5-(trifluoromethyl)phenyl]hydroxylamine
Formel: C₁₂H₁₆F₃N₃O
Molecular Weight: 275.27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04126038
CAS-Nummer: 866133-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR