8-(Trifluoromethyl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepin-2-one, tech

Lieferant: Apollo Scientific

PC28158-1G
APOSPC28158-1GEA 1570 EUR
APOSPC28158-1G
8-(Trifluoromethyl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepin-2-one, tech
8-(Trifluoromethyl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepin-2-one
Formel: C₁₀H₉F₃N₂O
Molecular Weight: 230,186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04126045
CAS-Nummer: 37040-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR