1,1,1-Trifluoro-3-{[(furan-2-yl)methyl]amino}propan-2-ol, tech
Lieferant: Apollo Scientific
PC28159-1G
APOSPC28159-1GEA
1570
EUR
APOSPC28159-1G
1,1,1-Trifluoro-3-{[(furan-2-yl)methyl]amino}propan-2-ol, tech
1,1,1-Trifluoro-3-{[(furan-2-yl)methyl]amino}propan-2-ol
Formel:
C₈H₁₀F₃NO₂ Molecular Weight: 209,166 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04123187 CAS-Nummer: 866135-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...