Ethyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperazine-1-carboxylate, tech

Lieferant: Apollo Scientific

PC28160-1G
APOSPC28160-1GEA 1570 EUR
APOSPC28160-1G
Ethyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperazine-1-carboxylate, tech
Ethyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperazine-1-carboxylate
Formel: C₁₀H₁₇F₃N₂O₃
Molecular Weight: 270,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05256288
CAS-Nummer: 866135-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR