3-{[(2-Bromophenyl)methyl]amino}-1,1,1-trifluoropropan-2-ol, tech

Lieferant: Apollo Scientific

PC28162-1G
APOSPC28162-1GEA 1570 EUR
APOSPC28162-1G
3-{[(2-Bromophenyl)methyl]amino}-1,1,1-trifluoropropan-2-ol, tech
3-{[(2-Bromophenyl)methyl]amino}-1,1,1-trifluoropropan-2-ol
Formel: C₁₀H₁₁BrF₃NO
Molecular Weight: 298,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05256310
CAS-Nummer: 866135-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR