4-Chloro-2-(pyridin-3-yl)-6-(trifluoromethyl)pyrimidine 95%
Lieferant: Apollo Scientific
PC28166-1G
APOSPC28166-1GEA
387
EUR
APOSPC28166-1G
4-Chloro-2-(pyridin-3-yl)-6-(trifluoromethyl)pyrimidine 95%
4-Chloro-2-(pyridin-3-yl)-6-(trifluoromethyl)pyrimidine
Formel:
C₁₀H₅ClF₃N₃ Molecular Weight: 259,615 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00140057 CAS-Nummer: 204394-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...