4-{[2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-1-yl]methyl}benzoic acid, tech
Lieferant: Apollo Scientific
PC28167-1G
APOSPC28167-1GEA
1570
EUR
APOSPC28167-1G
4-{[2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-1-yl]methyl}benzoic acid, tech
4-{[2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-1-yl]methyl}benzoic acid
Formel:
C₁₄H₁₀F₃NO₃ Molecular Weight: 297.229 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00140124 CAS-Nummer: 339024-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...