(E)-N'-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-N,N-dimethylmethanimidamide, tech

Lieferant: Apollo Scientific

PC28168-1G
APOSPC28168-1GEA 1570 EUR
APOSPC28168-1G
(E)-N'-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-N,N-dimethylmethanimidamide, tech
(E)-N'-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-N,N-dimethylmethanimidamide
Formel: C₉H₁₀ClF₃N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00215321
CAS-Nummer: 339029-44-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR