2-(2-Fluorophenyl)-2-(morpholin-4-yl)acetonitrile, tech

Lieferant: Apollo Scientific

PC28170-1G
APOSPC28170-1GEA 1570 EUR
APOSPC28170-1G
2-(2-Fluorophenyl)-2-(morpholin-4-yl)acetonitrile, tech
2-(2-Fluorophenyl)-2-(morpholin-4-yl)acetonitrile
Formel: C₁₂H₁₃FN₂O
Molecular Weight: 220,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173114
CAS-Nummer: 66549-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR