(1Z)-1-(Dimethylamino)-4-(4-fluorophenoxy)pent-1-en-3-one, tech
Lieferant: Apollo Scientific
PC28172-1G
APOSPC28172-1GEA
1570
EUR
APOSPC28172-1G
(1Z)-1-(Dimethylamino)-4-(4-fluorophenoxy)pent-1-en-3-one, tech
(1Z)-1-(Dimethylamino)-4-(4-fluorophenoxy)pent-1-en-3-one
Formel:
C₁₃H₁₆FNO₂ Molecular Weight: 237,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172162 CAS-Nummer: 1164512-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...