6-Methyl-3-[3-(trifluoromethyl)phenyl]-1,2,4-triazin-5-ol, tech

Lieferant: Apollo Scientific

PC28173-1G
APOSPC28173-1GEA 1570 EUR
APOSPC28173-1G
6-Methyl-3-[3-(trifluoromethyl)phenyl]-1,2,4-triazin-5-ol, tech
6-Methyl-3-[3-(trifluoromethyl)phenyl]-1,2,4-triazin-5-ol
Formel: C₁₁H₈F₃N₃O
Molecular Weight: 255,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665260
CAS-Nummer: 339102-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR