Methyl {[4-(trifluoromethyl)phenyl]carbamoyl}formate, tech

Lieferant: Apollo Scientific

PC28175-1G
APOSPC28175-1GEA 1570 EUR
APOSPC28175-1G
Methyl {[4-(trifluoromethyl)phenyl]carbamoyl}formate, tech
Methyl {[4-(trifluoromethyl)phenyl]carbamoyl}formate
Formel: C₁₀H₈F₃NO₃
Molecular Weight: 247,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568438
CAS-Nummer: 69066-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR