N-[(4-Fluorophenyl)methyl]methanesulphonamide, tech

Lieferant: Apollo Scientific

PC28176-1G
APOSPC28176-1GEA 1570 EUR
APOSPC28176-1G
N-[(4-Fluorophenyl)methyl]methanesulphonamide, tech
N-[(4-Fluorophenyl)methyl]methanesulphonamide
Formel: C₈H₁₀FNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01345415
CAS-Nummer: 339104-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR