3-{[(2-Chlorophenyl)methyl]amino}-1,1,1-trifluoropropan-2-ol, tech
Lieferant: Apollo Scientific
PC28177-1G
APOSPC28177-1GEA
1570
EUR
APOSPC28177-1G
3-{[(2-Chlorophenyl)methyl]amino}-1,1,1-trifluoropropan-2-ol, tech
3-{[(2-Chlorophenyl)methyl]amino}-1,1,1-trifluoropropan-2-ol
Formel:
C₁₀H₁₁ClF₃NO Molecular Weight: 253,649 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871719 CAS-Nummer: 478081-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...