1-(3,3,3-Trifluoro-2-hydroxypropyl)piperidin-4-ol, tech

Lieferant: Apollo Scientific

PC28178-1G
APOSPC28178-1GEA 1570 EUR
APOSPC28178-1G
1-(3,3,3-Trifluoro-2-hydroxypropyl)piperidin-4-ol, tech
1-(3,3,3-Trifluoro-2-hydroxypropyl)piperidin-4-ol
Formel: C₈H₁₄F₃NO₂
Molecular Weight: 213,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871721
CAS-Nummer: 478081-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR