1-[(4-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

PC28179-1G
APOSPC28179-1GEA 1570 EUR
APOSPC28179-1G
1-[(4-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
1-[(4-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₇H₁₁FN₂O₂
Molecular Weight: 294,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571450
CAS-Nummer: 478259-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR