6-Chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid, tech

Lieferant: Apollo Scientific

PC28182-1G
APOSPC28182-1GEA 1570 EUR
APOSPC28182-1G
6-Chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid, tech
6-Chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₉H₄ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06637318
CAS-Nummer: 874830-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR