3-Amino-2-(trifluoromethyl)-3,4-dihydroquinazolin-4-one, tech

Lieferant: Apollo Scientific

PC28186-1G
APOSPC28186-1GEA 1570 EUR
APOSPC28186-1G
3-Amino-2-(trifluoromethyl)-3,4-dihydroquinazolin-4-one, tech
3-Amino-2-(trifluoromethyl)-3,4-dihydroquinazolin-4-one
Formel: C₉H₆F₃N₃O
Molecular Weight: 229,159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00714352
CAS-Nummer: 152062-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR