Ethyl 4-(4-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech

Lieferant: Apollo Scientific

PC28187-1G
APOSPC28187-1GEA 1570 EUR
APOSPC28187-1G
Ethyl 4-(4-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Ethyl 4-(4-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate
Formel: C₁₅H₁₆FNO₃
Molecular Weight: 277,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975283
CAS-Nummer: 398995-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR