Methyl 4-(4-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech

Lieferant: Apollo Scientific

PC28188-1G
APOSPC28188-1GEA 1570 EUR
APOSPC28188-1G
Methyl 4-(4-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Methyl 4-(4-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate
Formel: C₁₄H₁₄FNO₃
Molecular Weight: 263,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00662364
CAS-Nummer: 299207-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR