Ethyl 4-(3,4-difluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Lieferant: Apollo Scientific
PC28189-1G
APOSPC28189-1GEA
1570
EUR
APOSPC28189-1G
Ethyl 4-(3,4-difluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Ethyl 4-(3,4-difluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate
Formel:
C₁₅H₁₅F₂NO₃ Molecular Weight: 295,281 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975288 CAS-Nummer: 478261-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...