1-Cyclopropyl-3-[(4-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

PC28191-1G
APOSPC28191-1GEA 1570 EUR
APOSPC28191-1G
1-Cyclopropyl-3-[(4-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one, tech
1-Cyclopropyl-3-[(4-fluorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one
Formel: C₁₅H₁₄FNO₂
Molecular Weight: 259,276 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871749
CAS-Nummer: 478247-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR