8-Chloro-3-(difluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech

Lieferant: Apollo Scientific

PC28192-1G
APOSPC28192-1GEA 1570 EUR
APOSPC28192-1G
8-Chloro-3-(difluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech
8-Chloro-3-(difluoromethyl)-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formel: C₈H₃ClF₅N₃
Molecular Weight: 271,575 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082839
CAS-Nummer: 478248-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR