1-[(3-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

PC28194-1G
APOSPC28194-1GEA 1570 EUR
APOSPC28194-1G
1-[(3-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
1-[(3-Fluorophenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₇H₁₁FN₂O₂
Molecular Weight: 294,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012366
CAS-Nummer: 439120-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR