(1E)-4,4,5,5,5-Pentafluoro-1-(piperidin-1-yl)pent-1-en-3-one, tech

Lieferant: Apollo Scientific

PC28196-1G
APOSPC28196-1GEA 1570 EUR
APOSPC28196-1G
(1E)-4,4,5,5,5-Pentafluoro-1-(piperidin-1-yl)pent-1-en-3-one, tech
(1E)-4,4,5,5,5-Pentafluoro-1-(piperidin-1-yl)pent-1-en-3-one
Formel: C₁₀H₁₂F₅NO
Molecular Weight: 257,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03102877
CAS-Nummer: 866144-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR