1-(4-Fluorobenzoyl)piperidin-4-one 95%

Lieferant: Apollo Scientific

PC28197-1G
APOSPC28197-1GEA 1120 EUR
APOSPC28197-1G
1-(4-Fluorobenzoyl)piperidin-4-one 95%
1-(4-Fluorobenzoyl)piperidin-4-one
Formel: C₁₂H₁₂FNO₂
Molecular Weight: 221,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115038
CAS-Nummer: 866144-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR