1-(4-Fluorophenyl)-4-(morpholin-4-yl)butan-1-one, tech
Lieferant: Apollo Scientific
PC28198-1G
APOSPC28198-1GEA
1570
EUR
APOSPC28198-1G
1-(4-Fluorophenyl)-4-(morpholin-4-yl)butan-1-one, tech
1-(4-Fluorophenyl)-4-(morpholin-4-yl)butan-1-one
Formel:
C₁₄H₁₈FNO₂ Molecular Weight: 251,297 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01737578 CAS-Nummer: 3800-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...