2-(4-Fluorophenyl)nicotinaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
2-(4-Fluorophenyl)pyridine-3-carbaldehyde
PC28199-1G
APOSPC28199-1GEA
671
EUR
APOSPC28199-1G
2-(4-Fluorophenyl)nicotinaldehyde 95%
2-(4-Fluorophenyl)nicotinaldehyde
Formel:
C₁₂H₈FNO Molecular Weight: 201,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659613 CAS-Nummer: 885950-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...