3-(4-Chlorophenyl)-4-fluoro-1H-pyrazol-5-amine 95%

Lieferant: Apollo Scientific

PC28204-1G
APOSPC28204-1GEA 538 EUR
APOSPC28204-1G
3-(4-Chlorophenyl)-4-fluoro-1H-pyrazol-5-amine 95%
3-(4-Chlorophenyl)-4-fluoro-1H-pyrazol-5-amine
Formel: C₉H₇ClFN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18426511
CAS-Nummer: 1246384-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR