4-Fluoro-3-(4-methylphenyl)-1H-pyrazol-5-amine 95%

Lieferant: Apollo Scientific

PC28207-1G
APOSPC28207-1GEA 538 EUR
APOSPC28207-1G
4-Fluoro-3-(4-methylphenyl)-1H-pyrazol-5-amine 95%
4-Fluoro-3-(4-methylphenyl)-1H-pyrazol-5-amine
Formel: C₁₀H₁₀FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18426516
CAS-Nummer: 1420660-49-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR