N-[(4-Fluoro-3-methoxyphenyl)methyl]cyclopentanamine, tech

Lieferant: Apollo Scientific

PC28210-1G
APOSPC28210-1GEA 1570 EUR
APOSPC28210-1G
N-[(4-Fluoro-3-methoxyphenyl)methyl]cyclopentanamine, tech
N-[(4-Fluoro-3-methoxyphenyl)methyl]cyclopentanamine
Formel: C₁₃H₁₈FNO
Molecular Weight: 223,287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21334157
CAS-Nummer: 1357147-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR