5-Chloro-1-(4-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde 95%
Lieferant: Apollo Scientific
PC28214-1G
APOSPC28214-1GEA
671
EUR
APOSPC28214-1G
5-Chloro-1-(4-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde 95%
5-Chloro-1-(4-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Formel:
C₁₃H₉ClFNO₂ Molecular Weight: 265,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10758060 CAS-Nummer: 1033463-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...