2-Chloro-N-(4-fluorophenyl)-N-(propan-2-yl)acetamide 95%

Lieferant: Apollo Scientific

PC28218-1G
APOSPC28218-1GEA 894 EUR
APOSPC28218-1G
2-Chloro-N-(4-fluorophenyl)-N-(propan-2-yl)acetamide 95%
2-Chloro-N-(4-fluorophenyl)-N-(propan-2-yl)acetamide
Formel: C₁₁H₁₃ClFNO
Molecular Weight: 229,678 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28506328
CAS-Nummer: 66602-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR