Ethyl 5-chloro-4-(trifluoromethyl)pyridine-2-carboxylate 97%
Lieferant: Apollo Scientific
PC28225-1G
APOSPC28225-1GEA
0
EUR
APOSPC28225-1G
Ethyl 5-chloro-4-(trifluoromethyl)pyridine-2-carboxylate 97%
Ethyl 5-chloro-4-(trifluoromethyl)pyridine-2-carboxylate
Formel:
C₉H₇ClF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22126112 CAS-Nummer: 1370587-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...