[5-Chloro-4-(trifluoromethyl)pyridin-2-yl]methanol 97%

Lieferant: Apollo Scientific

PC28226-1G
APOSPC28226-1GEA 1570 EUR
APOSPC28226-1G
[5-Chloro-4-(trifluoromethyl)pyridin-2-yl]methanol 97%
[5-Chloro-4-(trifluoromethyl)pyridin-2-yl]methanol
Formel: C₇H₅ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD22126113
CAS-Nummer: 1370587-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR