4-(Trifluoromethyl)benzene-1,2-diol 97%

Lieferant: Apollo Scientific

PC28227-1G
APOSPC28227-1GEA 538 EUR
APOSPC28227-1G
4-(Trifluoromethyl)benzene-1,2-diol 97%
4-(Trifluoromethyl)benzene-1,2-diol
Formel: C₇H₅F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18395730
CAS-Nummer: 37519-09-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR