3,3-Difluoro-2,3-dihydro-1H-indol-2-one, tech

Lieferant: Apollo Scientific

PC28228-1G
APOSPC28228-1GEA 671 EUR
APOSPC28228-1G
3,3-Difluoro-2,3-dihydro-1H-indol-2-one, tech
3,3-Difluoro-2,3-dihydro-1H-indol-2-one
Formel: C₈H₅F₂NO
Molecular Weight: 169,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18450082
CAS-Nummer: 197067-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR