1,3-Dimethyl-6-piperazino-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 1-[1,3-Dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-6-yl]piperazine

PC28231-1G
APOSPC28231-1GEA 671 EUR
APOSPC28231-1G
1,3-Dimethyl-6-piperazino-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 95%
1,3-Dimethyl-6-piperazino-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine
Formel: C₁₃H₁₆F₃N₅
Molecular Weight: 299.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09607951
CAS-Nummer: 952183-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR