2-[4-(Trifluoromethyl)phenethyl]piperazine 95%

Lieferant: Apollo Scientific

PC28232-1G
APOSPC28232-1GEA 671 EUR
APOSPC28232-1G
2-[4-(Trifluoromethyl)phenethyl]piperazine 95%
2-[4-(Trifluoromethyl)phenethyl]piperazine
Formel: C₁₃H₁₇F₃N₂
Molecular Weight: 258,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100209
CAS-Nummer: 1033463-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR