4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzaldehyde 97%

Lieferant: Apollo Scientific

PC28233-1G
APOSPC28233-1GEA 894 EUR
APOSPC28233-1G
4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzaldehyde 97%
4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzaldehyde
Formel: C₁₁H₇F₃N₂O
Molecular Weight: 240,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19586808
CAS-Nummer: 1393125-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR