4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid 97%

Lieferant: Apollo Scientific

PC28234-1G
APOSPC28234-1GEA 894 EUR
APOSPC28234-1G
4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid 97%
4-[4-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Formel: C₁₁H₇F₃N₂O₂
Molecular Weight: 256.181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19431019
CAS-Nummer: 1495970-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR