6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole

Lieferant: Apollo Scientific

Synonyme: 6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole

PC28238-5G
APOSPC28238-5GEA 140 EUR
APOSPC28238-5G
6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole
6-Bromo-4-fluoro-1-isopropyl-2-methyl-benzimidazole
Formel: C₁₁H₁₂BrFN₂
Molecular Weight: 271.129 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16660227
CAS-Nummer: 1231930-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR