Ethyl 5-(trifluoromethoxy)benzofuran-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC28249-250MG
APOSPC28249-250MGEA 87 EUR
APOSPC28249-250MG
Ethyl 5-(trifluoromethoxy)benzofuran-2-carboxylate
Ethyl 5-(trifluoromethoxy)benzofuran-2-carboxylate
Formel: C₁₂H₉F₃O₄
Molecular Weight: 274.193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28403629
CAS-Nummer: 896732-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR