5-(Trifluoromethoxy)benzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC28256-1G PC28256-250MG
APOSPC28256-1GEA 575 EUR
APOSPC28256-1G APOSPC28256-250MG
5-(Trifluoromethoxy)benzofuran-2-carboxylic acid
5-(Trifluoromethoxy)benzofuran-2-carboxylic acid
Formel: C₁₀H₅F₃O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD27925745
CAS-Nummer: 575469-37-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR