2,6-Dibrom-4-fluorophenylisocyanat 97%

Lieferant: Apollo Scientific
Achtung

PC28258-1G PC28258-25G PC28258-5G
APOSPC28258-1GEA 43.5 EUR
APOSPC28258-1G APOSPC28258-25G APOSPC28258-5G
2,6-Dibrom-4-fluorophenylisocyanat 97%
2,6-Dibrom-4-fluorophenylisocyanat

When melted, may contain small amount of sediment.

Formel: C₇H₂Br₂FNO
Lagertemperatur: Kühlschrank
MDL: MFCD00037041
CAS-Nummer: 76393-18-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR