2,6-Dibrom-4-fluorophenylisocyanat 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC28258-1G
PC28258-25G
PC28258-5G
APOSPC28258-1GEA
43.5
EUR
APOSPC28258-1G
APOSPC28258-25G
APOSPC28258-5G
2,6-Dibrom-4-fluorophenylisocyanat 97%
2,6-Dibrom-4-fluorophenylisocyanat
When melted, may contain small amount of sediment.
Formel:
C₇H₂Br₂FNO Lagertemperatur: Kühlschrank |
MDL:
MFCD00037041 CAS-Nummer: 76393-18-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...