3',5'-Difluor-2'-hydroxyacetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3,5-Difluoro-2-hydroxyphenyl)ethanone

PC2826-1G
APOSPC2826-1GEA 21.9 EUR
APOSPC2826-1G
3',5'-Difluor-2'-hydroxyacetophenon 98%
3',5'-Difluor-2'-hydroxyacetophenon
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
Schmelzpunkt: 105…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042476
CAS-Nummer: 140675-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR