8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC28270-1G PC28270-250MG
APOSPC28270-1GEA 383 EUR
APOSPC28270-1G APOSPC28270-250MG
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₉H₄ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12406878
CAS-Nummer: 1823188-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR